Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145047.4 | NP_659484.4 | 2 | 255 | ||
NM_206837.2 | NP_996668.1 | 2 | 255 | |||
XM_005270461.1 | XP_005270518.1 | 3 | 285 | |||
XM_005270462.1 | XP_005270519.1 | 3 | 285 | |||
XM_005270463.3 | XP_005270520.1 | 3 | 285 | |||
XM_011540680.2 | XP_011538982.1 | 4 | 327 | |||
XM_011540681.2 | XP_011538983.1 | 3 | 178 | |||
XM_017000303.1 | XP_016855792.1 | 2 | 191 | |||
XM_017000304.1 | XP_016855793.1 | 2 | 191 | |||
GenBank mRNA | AB079075.1 | - | 2 | 139 | ||
AF462348.1 | - | 2 | 297 | |||
AK098541.1 | - | 3 | 251 | |||
BC018069.1 | - | 2 | 221 | 4816622 | ||
BF691288.1 | - | 44 | ||||
BU854079.1 | - | 216 | ||||
DA297022.1 | - | 3 | 285 |