Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080738.3 | NP_542776.1 | 6 | 608 | ||
NM_145861.2 | NP_665860.2 | 6 | 489 | |||
GenBank mRNA | AK096339.1 | - | 6 | 608 | ||
AK290862.1 | - | 6 | 525 | |||
AK291930.1 | - | 6 | 523 | |||
AK314634.1 | - | 6 | 780 | |||
AY028913.1 | - | 6 | 489 | |||
AY028914.1 | - | 6 | 592 | |||
AY071862.1 | - | 6 | 394 | |||
BC114557.1 | - | 6 | 444 | |||
BC114560.1 | - | 6 | 444 | |||
BC128082.1 | - | 6 | 444 | 40006675 |