Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042552.2 | NP_001036017.1 | 3, 4 | 261 | ||
NM_001042553.2 | NP_001036018.1 | 3, 4 | 261 | |||
NM_001146169.1 | NP_001139641.1 | 3, 4 | 261 | |||
NM_001146170.1 | NP_001139642.1 | 3, 4 | 261 | |||
NM_001146171.1 | NP_001139643.1 | 3, 4 | 261 | |||
XM_011509202.2 | XP_011507504.1 | 3, 4 | 256 | |||
XM_017000328.1 | XP_016855817.1 | 3, 4 | 256 | |||
XM_017000329.1 | XP_016855818.1 | 3, 4 | 256 | |||
XM_017000330.1 | XP_016855819.1 | 3, 4 | 256 | |||
XM_017000331.1 | XP_016855820.1 | 3, 4 | 256 | |||
GenBank mRNA | AK293129.1 | - | 3, 4 | 261 | ||
AK295335.1 | - | 2, 3 | 186 | |||
AK304557.1 | - | 3, 4 | 219 | |||
AL832248.1 | - | 3, 4 | 203 | |||
BC048115.1 | - | 3, 4 | 172 | |||
BC113638.1 | - | 3, 4 | 205 | 8322690 | ||
BC117485.1 | - | 3, 4 | 205 | 40126036 | ||
BC143956.1 | - | 3, 4 | 205 | 9052474 | ||
BC143957.1 | - | 3, 4 | 205 |