Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s43376
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
4
449
NM_001283116.2
NP_001270045.1
3
203
NM_001283129.2
NP_001270058.1
3
443
NM_001283148.2
NP_001270077.1
3
203
NM_001283154.2
NP_001270083.1
3
315
NM_001283179.2
NP_001270108.1
3
330
NM_001283186.2
NP_001270115.1
3
203
NM_001283199.2
NP_001270128.1
3
315
NM_001283215.2
NP_001270144.1
3
443
NM_001283235.2
NP_001270164.1
3
203
NM_001283248.2
NP_001270177.1
3
203
NM_001322141.1
NP_001309070.1
3
443
NM_001322142.1
NP_001309071.1
3
203
NM_001322143.1
NP_001309072.1
3
516
NM_001322144.1
NP_001309073.1
3
443
NM_001322145.1
NP_001309074.1
3
516
NM_001322146.1
NP_001309075.1
3
276
NM_001322147.1
NP_001309076.1
3
443
NM_001322148.1
NP_001309077.1
3
443
NM_001322149.1
NP_001309078.1
3
443
NM_001322150.1
NP_001309079.1
3
443
NM_001322153.1
NP_001309082.1
3
203
NM_001322155.1
NP_001309084.1
3
276
NM_001322160.1
NP_001309089.1
3
328
NM_001322163.1
NP_001309092.1
4
449
NM_001322166.1
NP_001309095.1
3
276
NM_001322167.1
NP_001309096.1
3
328
NM_001322169.1
NP_001309098.1
3
203
NM_001322171.1
NP_001309100.1
3
276
NM_001322172.1
NP_001309101.1
3
568
NM_001322173.1
NP_001309102.1
3
443
NM_001322174.1
NP_001309103.1
3
330
NM_001322175.1
NP_001309104.1
3
328
NM_152906.6
NP_690870.3
3
315
NR_104274.2
3
203
NR_104275.2
3
443
NR_136206.1
3
276
NR_136211.1
3
276
NR_136212.1
3
276
XM_011529863.1
XP_011528165.1
3
277
XM_011529865.1
XP_011528167.1
2
219
XM_011529867.1
XP_011528169.1
4
413
XM_017028577.1
XP_016884066.1
3
276
XM_017028578.1
XP_016884067.1
3
295
XM_017028579.1
XP_016884068.1
3
324
XM_017028580.1
XP_016884069.1
4
413
XM_017028581.1
XP_016884070.1
3
330
XM_017028582.1
XP_016884071.1
4
413
XM_017028583.1
XP_016884072.1
3
160
XM_017028584.1
XP_016884073.1
3
423
XM_017028585.1
XP_016884074.1
3
275
XM_017028586.1
XP_016884075.1
3
200
XM_017028587.1
XP_016884076.1
3
330
XM_017028588.1
XP_016884077.1
3
256
GenBank mRNA
AA319727.1
-
3
171
AK092484.1
-
3
158
AK289862.1
-
4
418
AK295210.1
-
2
329
AK297075.1
-
3
160
AK298593.1
-
3
161
AK301054.1
-
3
162
AK301366.1
-
3
402
AK309398.1
-
2
83
AK311676.1
-
3
162
AK316020.1
-
3
164
AK316056.1
-
3
423
BC041339.1
-
3
278
5284126
BG685068.1
-
1
53
BI544307.1
-
3
278
BI911898.1
-
3
266
BM013784.1
-
3
486
BP193987.1
-
3
401
BP303570.1
-
2
100
BU173566.1
-
3
235
BX345246.2
-
3
315
CD675132.1
-
3
257
CF552364.1
-
2
174
CR456355.1
-
2
130
CR627452.1
-
3
158
DA176876.1
-
3
201
DC301369.1
-
3
172
DC303300.1
-
4
418
DC315148.1
-
3
161
DC318353.1
-
3
203
DC332363.1
-
3
423
DC349456.1
-
3
399
HY135099.1
-
3
330
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline