Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014208.3 | NP_055023.2 | 3944 | |||
GenBank mRNA | AF094508.1 | - | 2285 |