Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152384.2 | NP_689597.1 | 4 | 280 | ||
GenBank mRNA | AK312680.1 | - | 4 | 260 | ||
AL834305.1 | - | 4 | 271 | |||
AY604003.1 | - | 4 | 296 | |||
AY604004.1 | - | 4 | 296 | |||
BC044593.1 | - | 4 | 277 | 5272889 | ||
BP311042.1 | - | 4 | 281 | |||
KU178857.1 | - | 4 | 222 | |||
KU178858.1 | - | 4 | 222 |