Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004416.2 | NP_004407.2 | 4, 5 | 1493 | ||
XM_011538009.2 | XP_011536311.1 | 5, 6 | 2135 | |||
GenBank mRNA | AF053700.1 | - | 4, 5 | 1660 | ||
BC048216.1 | - | 4, 5 | 1435 | 5931062 |