Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039845.2 | NP_001034934.1 | 6 | 1228 | ||
NM_001282940.1 | NP_001269869.1 | 6 | 1228 | |||
XM_005246315.4 | XP_005246372.1 | 5 | 1049 | |||
XM_011510628.2 | XP_011508930.1 | 6 | 1214 | |||
XM_017003373.1 | XP_016858862.1 | 5 | 897 | |||
XM_017003374.1 | XP_016858863.1 | 5 | 884 | |||
GenBank mRNA | AK058070.1 | - | 5 | 858 | ||
AK292378.1 | - | 6 | 1226 | |||
AK292386.1 | - | 6 | 965 | |||
AK308257.1 | - | 6 | 1005 | |||
AK309144.1 | - | 5 | 892 | |||
BC088356.1 | - | 6 | 982 | 30383553 |