Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004416.2 | NP_004407.2 | 9 | 2092 | ||
XM_011538009.2 | XP_011536311.1 | 10 | 2734 | |||
GenBank mRNA | AF053700.1 | - | 9 | 2259 | ||
BC005816.2 | - | 5 | 571 | |||
BC048216.1 | - | 9 | 2034 | 5931062 |