Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301684.1 | NP_001288613.1 | 5 | 753 | ||
NM_001301685.1 | NP_001288614.1 | 7 | 1009 | |||
NM_138803.3 | NP_620158.3 | 7 | 1009 | |||
XM_005246319.3 | XP_005246376.1 | 8 | 896 | |||
XM_011510638.2 | XP_011508940.1 | 8 | 897 | |||
XM_017003392.1 | XP_016858881.1 | 7 | 1009 | |||
XM_017003393.1 | XP_016858882.1 | 7 | 692 | |||
XM_017003394.1 | XP_016858883.1 | 6 | 842 | |||
XM_017003395.1 | XP_016858884.1 | 6 | 905 | |||
XM_017003396.1 | XP_016858885.1 | 6 | 905 | |||
XM_017003397.1 | XP_016858886.1 | 4 | 382 | |||
GenBank mRNA | AF458588.1 | - | 7 | 958 | ||
AK058105.1 | - | 8 | 871 | |||
AK308783.1 | - | 5 | 753 | |||
AK309246.1 | - | 6 | 842 | |||
AK311042.1 | - | 5 | 783 | |||
BC015395.1 | - | 7 | 958 | |||
BC105928.1 | - | 5 | 566 | |||
BC107596.1 | - | 5 | 566 | 40025915 | ||
BC107597.1 | - | 7 | 822 | 40025920 |