Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175056.1 | NP_778226.1 | 5, 6 | 723 | ||
XM_017005703.1 | XP_016861192.1 | 6, 7 | 905 | |||
XM_017005704.1 | XP_016861193.1 | 5, 6 | 761 | |||
GenBank mRNA | AY090780.1 | - | 5, 6 | 723 | ||
BC031261.1 | - | 12, 13 | 1457 | 5273136 | ||
BC143817.1 | - | 6, 7 | 819 |