Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1FOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308262.1 | NP_001295191.1 | 3 | 493 | ||
NM_153833.2 | NP_722575.1 | 4 | 784 | |||
XM_017005733.1 | XP_016861222.1 | 3 | 530 | |||
GenBank mRNA | AY158091.1 | - | 4 | 784 | ||
BC047943.1 | - | 3 | 492 | 5742122 | ||
BM564157.1 | - | 510 |