Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144643.3 | NP_653244.2 | 20, 21 | 2513 | ||
XM_017007716.1 | XP_016863205.1 | 20, 21 | 2525 | |||
XM_017007719.1 | XP_016863208.1 | 20, 21 | 2046 | |||
XM_017007720.1 | XP_016863209.1 | 21, 22 | 2425 | |||
XM_017007721.1 | XP_016863210.1 | 14, 15 | 1627 | |||
GenBank mRNA | AK055217.1 | - | 20, 21 | 2513 | ||
AK093361.1 | - | 14, 15 | 1666 | |||
AK122852.1 | - | 10, 11 | 3138 | |||
BC121057.1 | - | 11, 12 | 1176 | |||
BC121058.1 | - | 7, 8 | 777 | |||
BC128051.1 | - | 11, 12 | 1175 | |||
CD679143.1 | - | 3, 4 | 278 |