Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPPLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009994.2 | NP_001009994.1 | 4 | 495 | ||
NR_103525.1 | 3 | 401 | ||||
GenBank mRNA | AK124105.1 | - | 1 | 3755 | ||
BC130460.1 | - | 4 | 460 | 40146491 | ||
BC132968.1 | - | 4 | 460 | 40146990 | ||
BM782014.1 | - | 3 | 354 |