Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138571.4 | NP_612638.3 | 2 | 478 | ||
GenBank mRNA | AJ420577.1 | - | 2 | 414 | ||
AK057688.1 | - | 2 | 408 | |||
AK309857.1 | - | 2 | 129 | |||
AY035387.1 | - | 2 | 403 | |||
AY035388.1 | - | 2 | 403 | |||
AY486460.1 | - | 2 | 215 | |||
AY486461.1 | - | 2 | 215 | |||
BC015732.1 | - | 2 | 412 | 4875139 | ||
BX647228.1 | - | 2 | 468 | |||
DA393891.1 | - | 2 | 478 |