Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDUFAF6 SIRNA Assays ›
Gene Symbol:
NDUFAF6
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases:
C8orf38
Chromosome Location:
Chr.8: 94895271 - 95118496 on Build GRCh38
UniGene ID:
Hs.729144
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44040
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDUFAF6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152416.3
NP_689629.2
5, 6
597
XM_005250789.2
XP_005250846.1
5, 6
608
XM_005250790.2
XP_005250847.1
6, 7
708
XM_005250791.2
XP_005250848.1
6, 7
674
XM_005250792.2
XP_005250849.1
6, 7
678
XM_005250793.2
XP_005250850.1
6, 7
591
XM_011516833.2
XP_011515135.1
10, 11
1817
XM_011516834.2
XP_011515136.1
9, 10
1778
XM_011516835.2
XP_011515137.1
9, 10
1036
XM_011516836.2
XP_011515138.1
9, 10
1719
XM_011516837.2
XP_011515139.1
10, 11
1777
XM_011516838.2
XP_011515140.1
9, 10
1654
XM_011516839.2
XP_011515141.1
8, 9
1556
XM_011516840.2
XP_011515142.1
9, 10
1650
XM_011516841.2
XP_011515143.1
9, 10
1679
XM_011516842.2
XP_011515144.1
8, 9
1552
XM_017013027.1
XP_016868516.1
9, 10
1028
XM_017013028.1
XP_016868517.1
8, 9
928
XM_017013029.1
XP_016868518.1
7, 8
1517
XM_017013030.1
XP_016868519.1
7, 8
1262
XM_017013031.1
XP_016868520.1
8, 9
1086
XM_017013032.1
XP_016868521.1
6, 7
708
XM_017013033.1
XP_016868522.1
5, 6
405
XM_017013034.1
XP_016868523.1
4, 5
378
XM_017013035.1
XP_016868524.1
8, 9
1130
XM_017013036.1
XP_016868525.1
8, 9
1462
XM_017013037.1
XP_016868526.1
6, 7
588
XM_017013038.1
XP_016868527.1
7, 8
657
XM_017013039.1
XP_016868528.1
6, 7
814
GenBank mRNA
AA625387.1
-
1, 2
61
AK298631.1
-
5, 6
517
AK301414.1
-
6, 7
639
AY444560.1
-
6, 7
668
BC028166.1
-
7, 8
930
BF030925.1
-
1, 2
109
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biosynthetic process
mitochondrial respiratory chain complex I assembly
transmembrane transport
transferase activity
transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0