Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144651.4 | NP_653252.3 | 15 | 1979 | ||
XM_006716420.3 | XP_006716483.1 | 13 | 3277 | |||
XM_011517456.2 | XP_011515758.1 | 14 | 3493 | |||
XM_011517457.2 | XP_011515759.1 | 14 | 3493 | |||
XM_011517458.2 | XP_011515760.1 | 13 | 1653 | |||
XM_011517459.2 | XP_011515761.1 | 15 | 3565 | |||
XM_017013041.1 | XP_016868530.1 | 12 | 1614 | |||
GenBank mRNA | AY877349.1 | - | 15 | 1979 | ||
EU170240.1 | - | 15 | 1878 | |||
KJ545488.1 | - | 13 | 1690 |