Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178819.3 | NP_848934.1 | 10, 11 | 1985 | ||
XM_011544392.2 | XP_011542694.1 | 11, 12 | 2108 | |||
XM_011544393.1 | XP_011542695.1 | 10, 11 | 1997 | |||
XM_011544394.1 | XP_011542696.1 | 10, 11 | 2034 | |||
XM_017013042.1 | XP_016868531.1 | 11, 12 | 2135 | |||
XM_017013043.1 | XP_016868532.1 | 11, 12 | 2006 | |||
GenBank mRNA | AF406612.1 | - | 10, 11 | 1973 | ||
AL834255.1 | - | 6, 7 | 470 | |||
AY358670.1 | - | 9, 10 | 1122 | |||
AY513610.1 | - | 9, 10 | 1205 | |||
BC051377.1 | - | 9, 10 | 1165 | 6649895 | ||
BC061884.1 | - | 9, 10 | 1165 | 6577569 |