Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138702.1 | NP_619647.1 | 7 | 1666 | ||
NM_177456.2 | NP_803251.1 | 5 | 1299 | |||
XM_011531267.2 | XP_011529569.1 | 3 | 1135 | |||
XM_017029265.1 | XP_016884754.1 | 7 | 2193 | |||
GenBank mRNA | AF490508.1 | - | 7 | 1666 | ||
BC103893.1 | - | 2 | 882 | 40005064 | ||
BC103894.1 | - | 3 | 692 |