Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144658.3 | NP_653259.3 | 25 | 2795 | ||
XM_005262368.3 | XP_005262425.1 | 25 | 3040 | |||
XM_005262369.4 | XP_005262426.1 | 25 | 2734 | |||
XM_011531276.2 | XP_011529578.1 | 25 | 2675 | |||
XM_011531278.2 | XP_011529580.1 | 12 | 1161 | |||
GenBank mRNA | AK124735.1 | - | 19 | 2180 | ||
AK125641.1 | - | 4 | 307 | |||
AY692226.1 | - | 25 | 2795 |