Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080874.3 | NP_543150.1 | 5 | 694 | ||
XM_005262759.1 | XP_005262816.1 | 7 | 1123 | |||
XM_011531616.2 | XP_011529918.1 | 5 | 802 | |||
XM_011531617.2 | XP_011529919.1 | 5 | 1385 | |||
GenBank mRNA | AK096441.1 | - | 5 | 596 | ||
AK098693.1 | - | 5 | 599 | |||
AY057053.1 | - | 5 | 580 | |||
BC065710.1 | - | 5 | 601 | 4272258 | ||
BX647857.1 | - | 5 | 695 |