Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF32 SIRNA Assays ›
Gene Symbol:
RNF32
Gene Name:
ring finger protein 32
Gene Aliases:
FKSG33, HSD15, LMBR2
Chromosome Location:
Chr.7: 156640286 - 156678333 on Build GRCh38
UniGene ID:
Hs.446194
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44297
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF32
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184996.1
NP_001171925.1
3
481
NM_001184997.1
NP_001171926.1
3
334
NM_001308273.1
NP_001295202.1
3
432
NM_001308274.1
NP_001295203.1
2
223
NM_030936.3
NP_112198.1
3
376
NR_131778.1
2
223
XM_005249522.4
XP_005249579.1
3
288
XM_011515804.2
XP_011514106.1
4
398
XM_011515805.2
XP_011514107.1
4
555
XM_011515806.2
XP_011514108.1
3
466
XM_011515807.2
XP_011514109.1
3
274
XM_011515808.2
XP_011514110.1
3
386
XM_011515809.2
XP_011514111.1
3
459
XM_011515810.2
XP_011514112.1
3
276
XM_011515811.2
XP_011514113.1
3
271
XM_011515812.2
XP_011514114.1
3
378
XM_011515813.2
XP_011514115.1
3
376
XM_017011753.1
XP_016867242.1
3
380
XM_017011754.1
XP_016867243.1
3
380
XM_017011755.1
XP_016867244.1
3
380
GenBank mRNA
AA412402.1
-
3
344
AF325690.1
-
3
284
AF441222.1
-
2
223
AF441224.1
-
2
223
AK307316.1
-
3
341
AK314067.1
-
3
481
AL136874.1
-
3
264
BC015416.1
-
2
234
4395421
BI460257.1
-
3
379
BI465216.1
-
3
312
BT007037.1
-
1
146
CR533513.1
-
1
146
DB198291.1
-
3
481
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0