Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103710.1 | 1, 2 | 401 | |||
GenBank mRNA | BX161496.1 | - | 1, 2 | 363 | ||
DA256249.1 | - | 1, 2 | 401 |