Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080879.2 | NP_543155.2 | 3 | 1257 | ||
XM_005262083.4 | XP_005262140.1 | 2 | 1734 | |||
XM_011530872.2 | XP_011529174.1 | 2 | 1731 | |||
GenBank mRNA | AF132748.1 | - | 1 | 980 | ||
AF422143.1 | - | 3 | 1260 | |||
AK056859.1 | - | 1 | 3034 |