Gene Symbol: VTI1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318203.1 | NP_001305132.1 | 7 | 851 | ||
NM_001318205.1 | NP_001305134.1 | 6 | 830 | |||
NM_145206.3 | NP_660207.2 | 6 | 830 | |||
XM_005269544.4 | XP_005269601.1 | 6 | 784 | |||
XM_005269545.4 | XP_005269602.1 | 7 | 851 | |||
XM_005269546.3 | XP_005269603.1 | 7 | 851 | |||
XM_005269547.4 | XP_005269604.1 | 6 | 784 | |||
XM_006717637.2 | XP_006717700.1 | 7 | 851 | |||
XM_011539328.2 | XP_011537630.1 | 7 | 851 | |||
XM_011539330.2 | XP_011537632.1 | 7 | 851 | |||
XM_011539331.2 | XP_011537633.1 | 7 | 851 | |||
XM_017015745.1 | XP_016871234.1 | 7 | 851 | |||
XM_017015746.1 | XP_016871235.1 | 6 | 784 | |||
XM_017015747.1 | XP_016871236.1 | 7 | 851 | |||
XM_017015748.1 | XP_016871237.1 | 6 | 784 | |||
GenBank mRNA | AI610960.1 | - | 2 | 116 | ||
AK303646.1 | - | 6 | 541 | |||
BC017052.1 | - | 6 | 560 | 3885564 | ||
BI909044.1 | - | 6 | 588 |