Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001269053.1 | NP_001255982.1 | 2 | 141 | ||
NM_138812.3 | NP_620167.1 | 3 | 264 | |||
XM_005269549.4 | XP_005269606.1 | 2 | 170 | |||
XM_005269550.4 | XP_005269607.1 | 2 | 497 | |||
XM_011539334.2 | XP_011537636.1 | 3 | 590 | |||
XM_011539335.1 | XP_011537637.1 | 3 | 590 | |||
XM_017015749.1 | XP_016871238.1 | 2 | 170 | |||
GenBank mRNA | AK125908.1 | - | 3 | 261 | ||
AK292542.1 | - | 2 | 497 | |||
BC019250.1 | - | 3 | 255 | 4828878 | ||
BI828082.1 | - | 2 | 136 |