Gene Symbol: HECTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284274.1 | NP_001271203.1 | 5 | 713 | ||
NM_173497.3 | NP_775768.3 | 5 | 713 | |||
NM_182765.4 | NP_877497.3 | 5 | 713 | |||
NR_104291.1 | 5 | 713 | ||||
XM_017015750.1 | XP_016871239.1 | 5 | 563 | |||
XM_017015751.1 | XP_016871240.1 | 5 | 659 | |||
GenBank mRNA | AK094625.1 | - | 5 | 654 | ||
AK295723.1 | - | 5 | 656 | |||
BC040187.1 | - | 5 | 656 | 4811444 | ||
BX537856.1 | - | 5 | 477 |