Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005744.3 | NP_005735.2 | 10, 11 | 1469 | ||
XM_011521438.2 | XP_011519740.1 | 9, 10 | 1445 | |||
XM_011521439.2 | XP_011519741.1 | 11, 12 | 983 | |||
GenBank mRNA | AB014774.1 | - | 1, 2 | 29 | ||
AB209432.1 | - | 11, 12 | 1098 | |||
AF072832.1 | - | 10, 11 | 1155 | |||
AJ009771.1 | - | 3, 4 | 265 | |||
AJ130976.1 | - | 10, 11 | 873 | |||
AJ243190.1 | - | 10, 11 | 1469 | |||
AK312715.1 | - | 10, 11 | 1468 | |||
BC051877.1 | - | 10, 11 | 1404 | 5742498 |