Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2ML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282424.2 | NP_001269353.1 | 12 | 1529 | ||
NM_144670.5 | NP_653271.2 | 23 | 3014 | |||
XM_011520566.2 | XP_011518868.1 | 23 | 2971 | |||
XM_011520567.2 | XP_011518869.1 | 23 | 2971 | |||
XM_017018868.1 | XP_016874357.1 | 23 | 2968 | |||
XM_017018869.1 | XP_016874358.1 | 23 | 2968 | |||
XM_017018870.1 | XP_016874359.1 | 23 | 2932 | |||
GenBank mRNA | AK123591.1 | - | 6 | 667 | ||
AK123592.1 | - | 12 | 1484 | |||
AK123601.1 | - | 20 | 2391 | |||
AK290355.1 | - | 23 | 2867 | |||
AK302555.1 | - | 12 | 1529 | |||
AL832139.1 | - | 23 | 2865 | |||
AL833479.1 | - | 21 | 2530 | |||
BX647674.1 | - | 5 | 469 | |||
BX648231.1 | - | 24 | 2866 |