Search Thermo Fisher Scientific
Gene Symbol: C12orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300940.1 | NP_001287869.1 | 2 | 474 | ||
NM_001300941.1 | NP_001287870.1 | 2 | 315 | |||
NM_152440.4 | NP_689653.3 | 2 | 474 | |||
GenBank mRNA | AK057111.1 | - | 2 | 441 | ||
AK129933.1 | - | 2 | 315 | |||
BC036246.1 | - | 2 | 443 | |||
BC071634.1 | - | 2 | 431 | 5182140 | ||
W44639.1 | - | 2 | 426 |