Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C12orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300940.1 | NP_001287869.1 | 3 | 1261 | ||
NM_001300941.1 | NP_001287870.1 | 3 | 1102 | |||
NM_152440.4 | NP_689653.3 | 3 | 1261 | |||
GenBank mRNA | AK057111.1 | - | 3 | 1228 | ||
AK129933.1 | - | 3 | 1102 | |||
BC036246.1 | - | 3 | 1230 | |||
BC071634.1 | - | 3 | 1218 | 5182140 |