Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194318.3 | NP_919299.3 | 15 | 1471 | ||
XM_006719768.3 | XP_006719831.1 | 16 | 1661 | |||
XM_011534936.1 | XP_011533238.1 | 14 | 1340 | |||
XM_011534938.2 | XP_011533240.1 | 14 | 1309 | |||
XM_017020395.1 | XP_016875884.1 | 16 | 1564 | |||
GenBank mRNA | AB101481.1 | - | 15 | 1361 | ||
AK094979.1 | - | 2 | 117 | |||
AK291273.1 | - | 15 | 1460 | |||
AY190526.1 | - | 15 | 1361 | |||
BC068595.1 | - | 15 | 1451 | 4837250 | ||
BX647932.1 | - | 2 | 537 |