Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIPOL1 SIRNA Assays ›
Gene Symbol:
MIPOL1
Gene Name:
mirror-image polydactyly 1
Gene Aliases:
CCDC193
Chromosome Location:
Chr.14: 37197888 - 37579207 on Build GRCh38
UniGene ID:
Hs.660396
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44720
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIPOL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195296.1
NP_001182225.1
6
859
NM_001195297.1
NP_001182226.1
6
845
NM_138731.6
NP_620059.1
5
706
XM_011536463.2
XP_011534765.1
8
1092
XM_011536467.2
XP_011534769.1
8
1089
XM_017020991.1
XP_016876480.1
8
1092
XM_017020992.1
XP_016876481.1
7
955
XM_017020993.1
XP_016876482.1
7
965
XM_017020994.1
XP_016876483.1
4
610
XM_017020995.1
XP_016876484.1
6
826
XM_017020996.1
XP_016876485.1
6
842
XM_017020997.1
XP_016876486.1
5
715
XM_017020998.1
XP_016876487.1
8
1091
XM_017020999.1
XP_016876488.1
9
1167
XM_017021000.1
XP_016876489.1
5
651
XM_017021001.1
XP_016876490.1
6
790
XM_017021002.1
XP_016876491.1
2
241
XM_017021003.1
XP_016876492.1
8
1090
XM_017021004.1
XP_016876493.1
4
618
XM_017021005.1
XP_016876494.1
5
713
XM_017021006.1
XP_016876495.1
8
1090
XM_017021007.1
XP_016876496.1
8
1090
XM_017021008.1
XP_016876497.1
5
649
XM_017021009.1
XP_016876498.1
6
762
XM_017021010.1
XP_016876499.1
2
241
XM_017021011.1
XP_016876500.1
4
574
XM_017021012.1
XP_016876501.1
6
762
XM_017021013.1
XP_016876502.1
2
241
XM_017021014.1
XP_016876503.1
8
1089
XM_017021015.1
XP_016876504.1
4
573
XM_017021016.1
XP_016876505.1
8
1089
XM_017021017.1
XP_016876506.1
8
1089
XM_017021018.1
XP_016876507.1
8
1089
GenBank mRNA
AK291850.1
-
6
770
AY059470.1
-
6
670
BC036124.1
-
6
731
5298587
BC036523.1
-
5
574
4828885
BX537396.1
-
5
669
BX537870.1
-
1
55
BX647795.1
-
5
545
CB105473.1
-
4
623
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0