Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIPOL1 SIRNA Assays ›
Gene Symbol:
MIPOL1
Gene Name:
mirror-image polydactyly 1
Gene Aliases:
CCDC193
Chromosome Location:
Chr.14: 37197888 - 37579207 on Build GRCh38
UniGene ID:
Hs.660396
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44721
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIPOL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195296.1
NP_001182225.1
11
1404
NM_001195297.1
NP_001182226.1
11
1390
NM_138731.6
NP_620059.1
10
1251
XM_011536463.2
XP_011534765.1
13
1637
XM_011536467.2
XP_011534769.1
13
1634
XM_017020991.1
XP_016876480.1
13
1637
XM_017020992.1
XP_016876481.1
12
1500
XM_017020993.1
XP_016876482.1
12
1510
XM_017020994.1
XP_016876483.1
9
1155
XM_017020995.1
XP_016876484.1
11
1371
XM_017020996.1
XP_016876485.1
11
1387
XM_017020997.1
XP_016876486.1
10
1260
XM_017020998.1
XP_016876487.1
13
1636
XM_017020999.1
XP_016876488.1
14
1712
XM_017021000.1
XP_016876489.1
10
1196
XM_017021001.1
XP_016876490.1
11
1335
XM_017021002.1
XP_016876491.1
7
786
XM_017021003.1
XP_016876492.1
13
1635
XM_017021004.1
XP_016876493.1
9
1163
XM_017021005.1
XP_016876494.1
10
1258
XM_017021006.1
XP_016876495.1
13
1635
XM_017021007.1
XP_016876496.1
13
1635
XM_017021008.1
XP_016876497.1
10
1194
XM_017021009.1
XP_016876498.1
11
1307
XM_017021010.1
XP_016876499.1
7
786
XM_017021011.1
XP_016876500.1
9
1119
XM_017021012.1
XP_016876501.1
11
1307
XM_017021013.1
XP_016876502.1
7
786
XM_017021014.1
XP_016876503.1
13
1634
XM_017021015.1
XP_016876504.1
9
1118
XM_017021016.1
XP_016876505.1
13
1634
XM_017021017.1
XP_016876506.1
13
1634
XM_017021018.1
XP_016876507.1
13
1634
XM_017021019.1
XP_016876508.1
3
4101
GenBank mRNA
AK291850.1
-
11
1315
AY059470.1
-
11
1215
BC035870.1
-
4
410
5245356
BC036124.1
-
11
1276
5298587
BC036523.1
-
10
1119
4828885
BX537396.1
-
10
1214
KU178867.1
-
3
205
KU178868.1
-
4
253
KU178869.1
-
3
205
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0