Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172365.2 | NP_758953.1 | 6, 7 | 529 | ||
XM_005267354.3 | XP_005267411.1 | 6, 7 | 449 | |||
XM_005267355.3 | XP_005267412.1 | 5, 6 | 376 | |||
XM_011536468.2 | XP_011534770.1 | 6, 7 | 497 | |||
GenBank mRNA | AK058044.1 | - | 6, 7 | 529 | ||
BC068989.1 | - | 6, 7 | 494 | 30389944 |