Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206673.1 | NP_001193602.1 | 9 | 773 | ||
NM_181533.3 | NP_853511.2 | 8 | 645 | |||
NM_181814.1 | NP_861535.1 | 7 | 542 | |||
XM_006720052.1 | XP_006720115.1 | 7 | 539 | |||
XM_011536472.1 | XP_011534774.1 | 9 | 843 | |||
XM_011536474.2 | XP_011534776.1 | 9 | 1294 | |||
XM_017021031.1 | XP_016876520.1 | 8 | 707 | |||
GenBank mRNA | AK128464.1 | - | 7 | 539 | ||
AY311396.1 | - | 9 | 773 | |||
AY311397.1 | - | 8 | 645 | |||
AY311398.1 | - | 7 | 542 | |||
AY311399.1 | - | 6 | 469 | |||
BC034603.1 | - | 7 | 509 | 4778051 | ||
BC107141.1 | - | 7 | 542 | 40030548 | ||
BC107142.1 | - | 7 | 540 | 40030549 |