Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289082.1 | NP_001276011.1 | 6 | 853 | ||
NM_001289083.1 | NP_001276012.1 | 7 | 838 | |||
NM_001289084.1 | NP_001276013.1 | 6 | 837 | |||
NM_001289085.1 | NP_001276014.1 | 7 | 966 | |||
NM_001289086.1 | NP_001276015.1 | 6 | 988 | |||
NM_001289087.1 | NP_001276016.1 | 5 | 769 | |||
NM_139018.4 | NP_620587.2 | 6 | 792 | |||
NR_110298.1 | 5 | 830 | ||||
XM_017024212.1 | XP_016879701.1 | 6 | 901 | |||
XM_017024213.1 | XP_016879702.1 | 7 | 963 | |||
XM_017024214.1 | XP_016879703.1 | 7 | 953 | |||
XM_017024215.1 | XP_016879704.1 | 7 | 847 | |||
XM_017024216.1 | XP_016879705.1 | 5 | 882 | |||
XM_017024217.1 | XP_016879706.1 | 5 | 868 | |||
GenBank mRNA | BM923728.1 | - | 3 | 130 |