Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321381.1 | NP_001308310.1 | 6 | 1481 | ||
NM_173079.3 | NP_775102.2 | 5 | 1475 | |||
XM_005257078.3 | XP_005257135.1 | 6 | 1536 | |||
XM_005257080.3 | XP_005257137.1 | 5 | 1530 | |||
GenBank mRNA | AL831813.1 | - | 5 | 1458 | ||
AY189284.1 | - | 6 | 1457 | |||
BC014873.1 | - | 5 | 1463 | |||
BC039247.1 | - | 5 | 1472 | 5271271 |