Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DHRS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144683.3 | NP_653284.2 | 5 | 1542 | ||
GenBank mRNA | AK296017.1 | - | 3 | 1193 | ||
AY358566.1 | - | 5 | 1542 | |||
BC015582.2 | - | 4 | 1532 | 4637504 | ||
BQ052759.1 | - | 1 | 362 |