Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002914.2 | NP_001002914.1 | 1 | 2099 | ||
GenBank mRNA | AK056227.1 | - | 1 | 1826 | ||
AY646650.1 | - | 1 | 1826 | |||
BC019929.1 | - | 1 | 1350 | 5115130 |