Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097610.2 | NP_001091079.1 | 2 | 181 | ||
XM_011535362.2 | XP_011533664.1 | 3 | 461 |
Gene Symbol: SCGB1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145651.2 | NP_663626.2 | 2 | 181 | ||
XM_005252804.3 | XP_005252861.2 | 3 | 461 | |||
GenBank mRNA | AY026938.1 | - | 2 | 181 | ||
BC069287.1 | - | 2 | 181 | 7262728 | ||
BC111427.1 | - | 2 | 180 | 40069184 | ||
BC111433.1 | - | 2 | 180 | 40069182 |