Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308238.1 | NP_001295167.1 | 4 | 474 | ||
NM_173505.3 | NP_775776.2 | 4 | 474 | |||
XM_011525831.2 | XP_011524133.1 | 3 | 2414 | |||
XM_011525832.2 | XP_011524134.1 | 3 | 2532 | |||
GenBank mRNA | AK057782.1 | - | 4 | 404 | ||
AK313667.1 | - | 4 | 392 | |||
BC030622.1 | - | 4 | 479 | 4818619 |