Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042474.2 | NP_001035939.1 | 5 | 747 | ||
NM_152477.4 | NP_689690.3 | 5 | 602 | |||
XM_011526512.2 | XP_011524814.1 | 5 | 815 | |||
XM_011526514.2 | XP_011524816.1 | 5 | 681 | |||
XM_017026341.1 | XP_016881830.1 | 6 | 1290 | |||
XM_017026342.1 | XP_016881831.1 | 4 | 510 | |||
XM_017026343.1 | XP_016881832.1 | 4 | 556 | |||
GenBank mRNA | AK057290.1 | - | 5 | 1334 | ||
AK094310.1 | - | 5 | 744 | |||
AK308212.1 | - | 4 | 520 | |||
BC068453.1 | - | 5 | 584 | 30343899 | ||
CB960081.1 | - | 5 | 611 |