Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280799.1 | NP_001267728.1 | 4, 5 | 293 | ||
NM_144698.4 | NP_653299.4 | 6, 7 | 563 | |||
GenBank mRNA | AK091120.1 | - | 6, 7 | 528 | ||
AK294308.1 | - | 4, 5 | 272 | |||
AK296861.1 | - | 5, 6 | 470 | |||
BC101618.1 | - | 6, 7 | 510 | 8069124 | ||
BC101620.1 | - | 6, 7 | 510 | 8069126 |