Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277155.2 | NP_001264084.1 | 3 | 543 | ||
NM_152490.4 | NP_689703.1 | 2 | 420 | |||
XM_006711749.3 | XP_006711812.1 | 2 | 362 | |||
XM_017000394.1 | XP_016855883.1 | 3 | 543 | |||
XM_017000395.1 | XP_016855884.1 | 3 | 496 | |||
GenBank mRNA | AK091497.1 | - | 2 | 193 | ||
AK130297.1 | - | 2 | 364 | |||
BC016974.1 | - | 3 | 484 | |||
BC029564.1 | - | 2 | 420 | 4826983 |