Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181644.4 | NP_857595.3 | 8, 9 | 1447 | ||
XM_011509229.2 | XP_011507531.1 | 7, 8 | 1186 | |||
GenBank mRNA | AK057733.1 | - | 4, 5 | 517 | ||
AK090831.1 | - | 2, 3 | 174 | |||
AK091896.1 | - | 8, 9 | 1388 | |||
AK295148.1 | - | 7, 8 | 1034 | |||
AK304109.1 | - | 7, 8 | 1252 | |||
AK304120.1 | - | 7, 8 | 1113 | |||
AL713676.1 | - | 6, 7 | 753 | |||
AY358107.1 | - | 6, 7 | 834 | |||
BC036549.2 | - | 8, 9 | 1397 | 5264261 |