- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: GCSAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281834.1 | NP_001268763.1 | 7 | 789 | ||
NM_001281835.1 | NP_001268764.1 | 6 | 673 | |||
NM_001281836.1 | NP_001268765.1 | 6 | 557 | |||
NM_001281837.1 | NP_001268766.1 | 6 | 538 | |||
NM_001281838.1 | NP_001268767.1 | 5 | 497 | |||
NM_001281853.1 | NP_001268782.1 | 7 | 642 | |||
NM_145278.4 | NP_660321.1 | 5 | 442 | |||
XM_017000399.1 | XP_016855888.1 | 6 | 557 | |||
GenBank mRNA | AK123798.1 | - | 5 | 353 | ||
AK124520.1 | - | 7 | 732 | |||
AK126682.1 | - | 6 | 611 | |||
AK307081.1 | - | 5 | 478 | |||
AK307082.1 | - | 6 | 523 | |||
AK311994.1 | - | 5 | 393 | |||
BC024174.1 | - | 5 | 342 | 4109064 | ||
DB095029.1 | - | 6 | 538 |