Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303477.1 | NP_001290406.1 | 4 | 491 | ||
NM_001303478.1 | NP_001290407.1 | 4 | 470 | |||
NM_152504.3 | NP_689717.2 | 3 | 400 | |||
XM_006723549.3 | XP_006723612.1 | 3 | 331 | |||
XM_011529177.2 | XP_011527479.1 | 3 | 387 | |||
XM_017027684.1 | XP_016883173.1 | 4 | 502 | |||
GenBank mRNA | AK057796.1 | - | 3 | 397 | ||
AK292708.1 | - | 3 | 386 | |||
BC035800.1 | - | 3 | 390 | 5734487 | ||
BU656896.1 | - | 4 | 444 | |||
CD694965.1 | - | 3 | 399 |