Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 4 | 773 | ||
NM_001284264.1 | NP_001271193.1 | 4 | 602 | |||
NM_001284265.1 | NP_001271194.1 | 5 | 553 | |||
NM_173510.3 | NP_775781.1 | 5 | 827 | |||
GenBank mRNA | AK091133.1 | - | 5 | 753 | ||
AK127794.1 | - | 1 | 1044 | |||
AK289516.1 | - | 5 | 825 | |||
AK295140.1 | - | 4 | 502 | |||
AK302798.1 | - | 5 | 556 | |||
AK302925.1 | - | 4 | 719 | |||
BC040488.1 | - | 5 | 782 | 5272169 | ||
BC053874.1 | - | 5 | 787 | 5531689 | ||
BC065549.1 | - | 5 | 789 | 6045091 | ||
CR456461.1 | - | 4 | 350 |