Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 3 | 615 | ||
NM_001284264.1 | NP_001271193.1 | 3 | 444 | |||
NM_001284265.1 | NP_001271194.1 | 4 | 395 | |||
NM_173510.3 | NP_775781.1 | 4 | 669 | |||
GenBank mRNA | AK091133.1 | - | 4 | 595 | ||
AK289516.1 | - | 4 | 667 | |||
AK295140.1 | - | 3 | 344 | |||
AK302798.1 | - | 4 | 398 | |||
AK302925.1 | - | 3 | 561 | |||
BC040488.1 | - | 4 | 624 | 5272169 | ||
BC053874.1 | - | 4 | 629 | 5531689 | ||
BC065549.1 | - | 4 | 631 | 6045091 | ||
CR456461.1 | - | 3 | 192 | |||
DB087275.1 | - | 4 | 397 |